SY3371475916 Synthonix Corporation

1-Benzylazetidin-3-ol

  • MDL number MFCD00963607
  • Cas Number : 54881-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/0168.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/0168.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart