SY3371475965 Synthonix Corporation

5-Bromo-2-hydroxynicotinonitrile

  • MDL number MFCD07363806
  • Cas Number : 405224-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/0432.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/0432.html
hstatements H302 , H315 , H318 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart