SY3371476028 Synthonix Corporation

tert-Butyl 7-oxa-3-aza-bicyclo[4.1.0]heptane-3-carboxylate

  • MDL number MFCD12407050
  • Cas Number : 161157-50-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/0710.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/0710.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart