SY3371476198 Synthonix Corporation

7-Bromo-2,4-dichlorothieno[3,2-d]pyrimidine

  • MDL number MFCD11869289
  • Cas Number : 41102-25-4
  • Molecular Weight : 283.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/1471.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/1471.html
pictograms GHS06, GHS07, GHS08, GHS09
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart