SY3371476200 Synthonix Corporation

1-Bromo-3-ethylsulfonylbenzene

  • MDL number MFCD19982772
  • Cas Number : 153435-82-6
  • Molecular Weight : 249.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Viscous
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/1476.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/1476.html
pictograms GHS07
hstatements H301, H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart