SY3371476202 Synthonix Corporation

4-Benzyloxy-1-tert-butoxycarbonylindole-2-boronic acid

  • MDL number MFCD04973739
  • Cas Number : 850568-52-4
  • Molecular Weight : 367.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(benzyloxy)-1-(tert-butoxycarbonyl)-1H-indol-2-ylboronic acid
Inchi Code 1S/C20H22BNO5/c1-20(2,3)27-19(23)22-16-10-7-11-17(15(16)12-18(22)21(24)25)26-13-14-8-5-4-6-9-14/h4-12,24-25H,13H2,1-3H3
Inchi Key YXJBIADEQVCONT-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/1495.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/1495.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart