SY3371476210 Synthonix Corporation

5-Bromo-1H-imidazo[4,5-b]pyridine

  • MDL number MFCD11227145
  • Cas Number : 28279-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/1551.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/1551.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart