SY3371476256 Synthonix Corporation

5-Bromobenzo[B]thiophene

  • MDL number MFCD03069318
  • Cas Number : 4923-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/1871.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/1871.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart