SY3371476273 Synthonix Corporation

3-N-(BOC)aminomethylphenylboronic acid

  • MDL number MFCD06246052
  • Cas Number : 199609-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/1972.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/1972.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart