SY3371476281 Synthonix Corporation

trans-1-Boc-3-fluoro-4-hydroxypiperidine

  • MDL number MFCD22123259
  • Cas Number : 955029-44-4
  • Molecular Weight : 219.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (3R,4R)-3-fluoro-4-hydroxy-1-piperidinecarboxylate
Inchi Code 1S/C10H18FNO3/c1-10(2,3)15-9(14)12-5-4-8(13)7(11)6-12/h7-8,13H,4-6H2,1-3H3/t7-,8-/m1/s1
Inchi Key XRNLYXKYODGLMI-HTQZYQBOSA-N

Description

Purity 97.00%
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/2002.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/2002.html
pictograms GHS05, GHS07
hstatements H302, H315, H318, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart