SY3371476292 Synthonix Corporation

1-Benzyl-2-(chloromethyl)-1H -imidazole hydrochloride

  • MDL number MFCD00035210
  • Cas Number : 19276-03-0
  • Molecular Weight : 243.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/2089.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/2089.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart