SY3371476312 Synthonix Corporation

4-Bromo-2-ethoxythiazole

  • MDL number MFCD07787375
  • Cas Number : 240816-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/2261.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/2261.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart