SY3371476355 Synthonix Corporation

5-Bromo-2-methylsulphonylpyridine

  • MDL number MFCD04974510
  • Cas Number : 98626-95-0
  • Molecular Weight : 236.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/2647.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/2647.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart