SY3371476356 Synthonix Corporation

6-Bromo-1-(phenylsulfonyl)indole

  • MDL number MFCD03840610
  • Cas Number : 679794-03-7
  • Molecular Weight : 336.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromo-1-(phenylsulfonyl)-1H-indole
Inchi Code 1S/C14H10BrNO2S/c15-12-7-6-11-8-9-16(14(11)10-12)19(17,18)13-4-2-1-3-5-13/h1-10H
Inchi Key NENPDYQYNRFWQU-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/2691.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/2691.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart