SY3371476385 Synthonix Corporation

3-Bromo-2-fluoropyridine-4-boronic acid pinacol ester

  • MDL number MFCD08063110
  • Cas Number : 1150561-78-6
  • Molecular Weight : 301.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/2930.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/2930.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart