SY3371476421 Synthonix Corporation

6-Bromo-3-(methylthio)-[1,2,4]triazolo[4,3-a]pyridine

  • MDL number MFCD09475863
  • Cas Number : 941294-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/3324.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/3324.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart