SY3371476450 Synthonix Corporation

N-(BOC)-6-fluoroindole-2-boronic acid

  • MDL number MFCD09953519
  • Cas Number : 1000068-26-7
  • Molecular Weight : 279.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/3591.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/3591.html
pictograms GHS05, GHS07
hstatements H302, H315, H318, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart