SY3371476468 Synthonix Corporation

tert-Butyl 3-bromo-5,6-dihydro-[1,2,4]triazolo[4,3-a]pyrazine-7(8H)-carboxylate

  • Cas Number : 723286-80-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/4044.html

Safety Information

MSDS https://synthonix.com/4044.html
Availability details for
Quantity
  • Availability
  • Add to Cart