SY3371476472 Synthonix Corporation

1-(2,2-Diethoxyethyl)-5-nitro-1H-pyrrole-2-carboxylic acid ethyl ester

  • MDL number MFCD11973836
  • Cas Number : 1105187-49-2
  • Molecular Weight : 300.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/4066.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/4066.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart