SY3371476486 Synthonix Corporation

6-Bromo-3,4-dihydro-4,4-dimethylquinolin-2(1H)-one

  • MDL number MFCD00795100
  • Cas Number : 135631-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/4234.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/4234.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart