SY3371476491 Synthonix Corporation

7-Benzyl-5,6,7,8-tetrahydro-3H-pyrido[3,4-d]pyrimidin-4-one

  • MDL number MFCD09030048
  • Cas Number : 62458-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/4275.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/4275.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart