SY3371476495 Synthonix Corporation

tert-Butyl 2-bromo-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-carboxylate

  • Cas Number : 949922-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/4291.html

Safety Information

MSDS https://synthonix.com/4291.html
Availability details for
Quantity
  • Availability
  • Add to Cart