SY3371476497 Synthonix Corporation

6-(tert-Butoxycarbonyl-)-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-2-carboxylic acid

  • MDL number MFCD09878719
  • Cas Number : 165947-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/4297.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/4297.html
hstatements H315 , H317 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart