SY3371476498 Synthonix Corporation

tert-Butyl 7,8-dihydro-2-(methylsulfonyl)pyrido[4,3-d]pyrimidine-6(5H)-carboxylate

  • MDL number MFCD09839783
  • Cas Number : 259809-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/4301.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/4301.html
hstatements H301 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart