SY3371476563 Synthonix Corporation

7-Bromoindazole-1-carboxylic acid tert-butyl ester

  • MDL number MFCD11505878
  • Cas Number : 1092352-37-8
  • Molecular Weight : 297.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/4906.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/4906.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart