SY3371476594 Synthonix Corporation

6-Bromo-7-methyl[1,2,4]triazolo[1,5-a]pyridine

  • MDL number MFCD12026325
  • Cas Number : 1172534-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/5081.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/5081.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart