SY3371476639 Synthonix Corporation

6-Bromoimidazo[1,2-a]pyridin-8-amine

  • Cas Number : 676371-00-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/5290.html

Safety Information

MSDS https://synthonix.com/5290.html
Availability details for
Quantity
  • Availability
  • Add to Cart