SY3371476649 Synthonix Corporation

tert-Butyl 2-amino-5H-pyrrolo[3,4-d]pyrimidine-6(7H)-carboxylate

  • Cas Number : 1105187-42-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/5340.html

Safety Information

MSDS https://synthonix.com/5340.html
Availability details for
Quantity
  • Availability
  • Add to Cart