SY3371476650 Synthonix Corporation

7-Bromofuro[3,2-c]pyridin-4(5H)-one

  • Cas Number : 603301-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/5342.html

Safety Information

MSDS https://synthonix.com/5342.html
Availability details for
Quantity
  • Availability
  • Add to Cart