SY3371476672 Synthonix Corporation

2-(n-Butoxy)-6-methoxyphenylboronic acid pinacol ester

  • MDL number MFCD13152003
  • Cas Number : 1204580-83-5
  • Molecular Weight : 306.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/5452.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/5452.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart