SY3371476733 Synthonix Corporation

(3-(Bromomethyl)oxetan-3-yl)methanol

  • MDL number MFCD09800598
  • Cas Number : 22633-44-9
  • Molecular Weight : 181.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/5888.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/5888.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart