SY3371476736 Synthonix Corporation

1-tert-Butylsulfinyl-5-oxa-1-azaspiro[2.3]hexane

  • MDL number MFCD18325167
  • Cas Number : 1211284-25-1
  • Molecular Weight : 189.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/5964.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/5964.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart