SY3371476761 Synthonix Corporation

1-Benzhydryl-3-(trifluoromethyl)azetidin-3-ol

  • MDL number MFCD18839240
  • Cas Number : 848192-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://synthonix.com/6194.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6194.html
hstatements H302 , H315 , H318 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart