SY3371476824 Synthonix Corporation

tert-Butyl (1S,5R)-6-amino-3-azabicyclo[3.2.0]heptane-3-carboxylate

  • MDL number MFCD14581158
  • Cas Number : 1250884-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/6578.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/6578.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart