SY3371476832 Synthonix Corporation

Benzo[1,2-b:4,5-b’]dithiophene

  • MDL number MFCD14708168
  • Cas Number : 267-65-2
  • Molecular Weight : 190.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/6609.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/6609.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart