SY3371476929 Synthonix Corporation

6-Benzyl-5,6,7,8-tetrahydro-1,6-naphthyridin-3-amine

  • MDL number MFCD09025775
  • Cas Number : 214699-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/4581.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/4581.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart