SY3371476930 Synthonix Corporation

tert-Butyl 5,6-dihydroimidazo[1,2-a]pyrazine-7(8H)-carboxylate

  • MDL number MFCD09878590
  • Cas Number : 345311-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/4582.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/4582.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart