SY3371476957 Synthonix Corporation

4-Chloro-7H-pyrrolo[2,3-d]pyrimidine-6-carboxylic acid methyl ester

  • MDL number MFCD09702013
  • Cas Number : 944709-69-7
  • Molecular Weight : 211.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/0218.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/0218.html
pictograms GHS06, GHS07, GHS08, GHS09
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart