SY3371476980 Synthonix Corporation

5-Chloro-1H-pyrrolo[2,3-b]pyridin-2(3H)-one

  • MDL number MFCD12962791
  • Cas Number : 1190314-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/0439.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/0439.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart