SY3371476989 Synthonix Corporation

5-Chloro-1H-pyrrolo[2,3-c]pyridin-2(3H)-one

  • MDL number MFCD11518942
  • Cas Number : 136888-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/0457.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/0457.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart