SY3371477049 Synthonix Corporation

4-Chloro-1,7-naphthyridine

  • MDL number MFCD18382494
  • Cas Number : 16287-97-1
  • Molecular Weight : 164.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/1826.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/1826.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart