SY3371477052 Synthonix Corporation

2-Chloro-5-methylthiazole

  • MDL number MFCD03265390
  • Cas Number : 33342-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://synthonix.com/1978.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/1978.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart