SY3371477112 Synthonix Corporation

2-Chloro-4-(hydroxymethyl)thiazole

  • MDL number MFCD09265529
  • Cas Number : 5198-85-6
  • Molecular Weight : 149.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/2782.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/2782.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart