SY3371477117 Synthonix Corporation

3-Chloro-4-isopropoxycyclobutene-1,2-dione

  • MDL number MFCD09763707
  • Cas Number : 130837-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/2841.html

Safety Information

pcodes P280 ,
MSDS https://synthonix.com/2841.html
hstatements H314 ,
Availability details for
Quantity
  • Availability
  • Add to Cart