SY3371477126 Synthonix Corporation

2-Cyano-5-iodopyridine

  • MDL number MFCD10699132
  • Cas Number : 41960-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/3011.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/3011.html
hstatements H302 , H315 , H318 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart