SY3371477162 Synthonix Corporation

6-Chloro-4-cyclopropyl-1H-quinazolin-2-one

  • MDL number MFCD11109434
  • Cas Number : 150878-36-7
  • Molecular Weight : 220.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/3622.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/3622.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart