SY3371477222 Synthonix Corporation

2-Chloro-5-iodo-1H-benzo[d]imidazole

  • Cas Number : 256518-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/5322.html

Safety Information

MSDS https://synthonix.com/5322.html
Availability details for
Quantity
  • Availability
  • Add to Cart