SY3371477229 Synthonix Corporation

4-Chloro-5-fluoro-2-pyridinemethanol

  • MDL number MFCD09953777
  • Cas Number : 113209-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/5396.html

Safety Information

MSDS https://synthonix.com/5396.html
Availability details for
Quantity
  • Availability
  • Add to Cart