SY3371477230 Synthonix Corporation

1-Chloroisoquinoline-5-carboxylic acid hydrochloride

  • MDL number MFCD11518953
  • Cas Number : 223671-71-4
  • Molecular Weight : 244.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/5414.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/5414.html
pictograms GHS07
hstatements H302, H315, H317, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart