SY3371477231 Synthonix Corporation

2-Cyclopropylmethoxy-6-methoxyphenylboronic acid pinacol ester

  • MDL number MFCD13152001
  • Cas Number : 1204580-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/5450.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/5450.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart